Chromium standard solution, 1000 mg/L Cr in 0,5 M nitric acid (from Cr(NO₃)₃.9H₂O) standard for AAS
Catalog # KRAF03636.2000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:0,5 M HNO₃
- Pack type:Plastic bottle
- Pk:500 mL
- Element:Chromium
- Symbol:Cr
Specifications
About this item
Certifications: Traceable to NIST