Mangan 1000 mg/L v 0,5 M kyselině dusičné (from Mn(NO₃)₂.H₂O) standard pro ICP
Catalog # KRAF03822.0000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:0,5 M HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Manganese
- Symbol:Mn
Specifications
About this item
Certifications: Traceable to NIST