Order Entry
Germany
ContactUsLinkComponent
Ammonium standard solution, 1000 mg/L NH₄ in water standard for ion selective electrodes
Ammonium standard solution, 1000 mg/L NH₄ in water standard for ion selective electrodes
  REGEISENH45
 :  REAGECON
 :  
Ammonium standard solution, 1000 mg/L NH₄ in water standard for ion selective electrodes
  REGEISENH45
 :  REAGECON
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    Ammonium standard solution