About this item
Stand model, volume range 0,01 to 2500 ml. The PT 4000 will provide high dispersing tool tip speeds due to the motor speed of up to 40000 rpm. This high tip speed will provide small droplet or particle sizes for already proven applications and provides a means to produce nano particles. The motor, which is a brushless, wear-free (3-phase) high-frequency motor, has a low noise level at maximum speed making it ideal for use in a crowded laboratory environment.
- Aggregate tip speeds up to 40 m/s produce the finest particles possible
- Soft start prevents splashing of homogenate from sample vessel
- Thermal fuse for overload protection
- Speed remains constant even when solution viscosity changes
- Aggregate replacement using one-handed quick release coupling
The PT 4000 will provide high dispersing tool tip speeds due to the motor speed of up to 40000 rpm. This high tip speed will provide small droplet or particle sizes for already proven applications and provides a means to produce nano particles. The motor, which is a brushless, wear-free (3-phase) high-frequency motor, has a low noise level at maximum speed making it ideal for use in a crowded laboratory environment.