ContactUsLinkComponent
Petri dish safety dispenser
Petri dish safety dispenser
Catalog # MBVA00.1000.01
Supplier:  MBV
undefined
Petri dish safety dispenser
Catalog # MBVA00.1000.01
Supplier:  MBV
Specifications
  • Description:
    Petri dish dispenser, PSD-10
  • Pk:
    10
Specifications
About this item
PP dispenser for Petri dishes and RODAC plates.
  • Stackable
  • Absolute safety during transport
  • Autoclavable
The PSD-10 can be used in all microbiological laboratories. It serves to prevent lids accidental falling off and so protects the user and the environment from microorganisms and potentially toxic and dangerous contamination. It offers a safe way to transport Petri dishes and RODAC plates. Each Petri dish is individually packed in a container. The lid of the dish is pressed against the bottom part and thus it is impossible for the lid to fall off. By pressing on the colour clip, the plate can easily be removed by hand.
Dimensions of the container: 103×100×21 mm
Weight: ~38 g