Detergents for glassware and plastics, Aquet®, SP Bel-Art
148-1170
- Capacity:3,8 l
- Description:Aquet®, phosphate-free, neutral pH detergent concentrate
- Packed:3.8 litre plastic bottle comes with 473 ml working solution dispenser bottle.
- Pk:1
Phosphate-Free Liquid Detergent with Neutral pH
Safe for glassware, plastics and surfaces, this phosphate-free, nonionic surfactant will not etch or mar precision glassware. It rinses quickly and dries without wiping.