Order Entry
Germany
ContactUsLinkComponent
Sulphuric acid ≥78%, VOLUSOL® volumetric solution
Corrosive
Sulphuric acid ≥78%, VOLUSOL® volumetric solution
Catalog # VWRS83994.330
Supplier:  VWR Collection
CAS Number:  
Corrosive
Sulphuric acid ≥78%, VOLUSOL® volumetric solution
Catalog # VWRS83994.330
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :98.08 g/mol
Storage Temperature:Ambient
EINECS:231-639-5
UN:1830
ADR:8,II
 :GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589