Orders LinkContactUsLinkComponent
Rubber protection mats
Rubber protection mats
Supplier:   DEUTSCH NEUMANN
Rubber protection mats
Supplier:  DEUTSCH NEUMANN
Certificates
Rubber protection mats
Rubber protection mats
10 Options Available Below
Product Details & Documents
Black protection sheets made from fluoroelastomer rubber (FPM). Durometer 75 Shore A.
  • For temperature ranges from −10 to over +200 °C
  • Good resistance to ozone and weathering
  • Benzine and mineral oil resistant
Product Family Options
Product Information
  • Colour
    L×W
    Thickness
    Availability
    Price
  • Black
    200×200 mm
    1,00 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 140,00 €
    140,00 €
    1 * 1 ST
       140,00 €
  • Black
    200×200 mm
    1,50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 186,00 €
    186,00 €
    1 * 1 ST
       186,00 €
  • Black
    200×200 mm
    2,00 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 215,00 €
    215,00 €
    1 * 1 ST
       215,00 €
  • Black
    200×200 mm
    3,00 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 247,00 €
    247,00 €
    1 * 1 ST
       247,00 €
  • Black
    200×200 mm
    4,00 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 410,00 €
    410,00 €
    1 * 1 ST
       410,00 €
  • Black
    300×300 mm
    1,00 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 245,00 €
    245,00 €
    1 * 1 ST
       245,00 €
  • Black
    300×300 mm
    1,50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 252,00 €
    252,00 €
    1 * 1 ST
       252,00 €
  • Black
    300×300 mm
    2,00 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 326,00 €
    326,00 €
    1 * 1 ST
       326,00 €
  • Black
    300×300 mm
    3,00 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 401,00 €
    401,00 €
    1 * 1 ST
       401,00 €
  • Black
    300×300 mm
    4,00 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 550,00 €
    550,00 €
    1 * 1 ST
       550,00 €
This product has been blocked by your organization. Please contact your purchasing department for more information