Orders LinkContactUsLinkComponent
Surface protection mats
Surface protection mats
Supplier:   DEUTSCH NEUMANN
Surface protection mats
Supplier:  DEUTSCH NEUMANN
Certificates
Surface protection mats
Surface protection mats
5 Options Available Below
Product Details & Documents
Translucent bench mats made from silicone rubber (VMQ).
  • Temperature range from −60 to +200° C
  • Short periods up to 260° C
  • Durometer 60 Shore A
Product Family Options
Product Information
  • Colour
    L×W
    Thickness
    Availability
    Price
  • Translucent
    550×600 mm
    1,00 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 343,00 €
    343,00 €
    1 * 1 ST
       343,00 €
  • Translucent
    550×600 mm
    2,00 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 487,00 €
    487,00 €
    1 * 1 ST
       487,00 €
  • Translucent
    550×600 mm
    3,00 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 670,00 €
    670,00 €
    1 * 1 ST
       670,00 €
  • Translucent
    550×600 mm
    4,00 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 720,00 €
    720,00 €
    1 * 1 ST
       720,00 €
  • Translucent
    550×600 mm
    5,00 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 794,00 €
    794,00 €
    1 * 1 ST
       794,00 €