ContactUsLinkComponent
Tray, for Petri dish incubation, SP Bel-Art
Tray, for Petri dish incubation, SP Bel-Art
Catalog # BELAF189830000
undefined
Tray, for Petri dish incubation, SP Bel-Art
Catalog # BELAF189830000
Specifications
  • W×D×H:
    349×137×22 mm
  • Pk:
    1
Specifications
About this item
Tray made of white epoxy-coated steel.
  • Excellent aid for keeping cell cultures organised
  • With 29 holes of 19 mm diameter in bottom allow air passage
  • Holds 100 mm petri dishes securely on an incubator rack