ContactUsLinkComponent
SP Bel-Art sidENTRY™, Glove box
SP Bel-Art sidENTRY™, Glove box
Catalog # BELAH500260000
undefined
SP Bel-Art sidENTRY™, Glove box
Catalog # BELAH500260000
Specifications
  • Description:
    Glove box, acrylic, with two circular front glove ports (Ø 203,2 mm) and one large gasketed side door (44,5 cm²)
  • W×D×H ext.:
    762×610×610 mm
  • Pk:
    1
Specifications
About this item
This small and portable glove box is designed for remote and field use. It is manufactured from transparent acrylic plastic (6,35 mm thick).
  • Glove box stability is assured by four adjustable levelers that also level the box and all its contents
  • Stopcocks are included ready for assembly
  • Includes large gasketed door on side of unit
  • Features two 20,32 cm glove ports compatible with standard glove box gloves