Order Entry
Germany
ContactUsLinkComponent
Pen-Ray® Photochemical Quartz UV Lamps
Pen-Ray® Photochemical Quartz UV Lamps
Catalog # UVPA90-0049-01
undefined
Pen-Ray® Photochemical Quartz UV Lamps
Catalog # UVPA90-0049-01

Specifications

  • Description:
    Photochemical quartz lamp, lighted length of 63,5 mm
  • Seq:
    01
  • Pk:
    1
  • Wavelength:
    254 nm

Specifications

About this item