Order Entry
Germany
ContactUsLinkComponent
Desiccator cabinets, with gas ports, dry-keeper™, SP Bel-Art
Desiccator cabinets, with gas ports, dry-keeper™, SP Bel-Art
Catalog # BELAH420530002
undefined
Desiccator cabinets, with gas ports, dry-keeper™, SP Bel-Art
Catalog # BELAH420530002

Specifications

  • W×D×H ext.:
    210×270×170 mm
  • Description:
    Desiccator, clear polystyrene, 4 l
  • Pk:
    1
  • With:
    1 chamber, with 2 gas ports

Specifications

About this item

Compact clear polystyrene desiccators, up to 5 units can be safely stacked for maximum use of laboratory bench space.

  • Pull down door slides out of the way into a compartment beneath the cabinet
  • Rear mounted gas ports are tapered to accept tubing with 6,3 to 9,5 mm
  • Gas ports can be closed to use desiccator with standard desiccant cartridges