Goggles, uvex 9405
111-2613
: UVEX
- Type:Anti-UV, anti-fog
- Colour:Translucent grey
- Lenses:Clear cellulose acetate
- Pk:1
The large goggles are suitable to use with prescription eye wear and filtering half masks.
- Soft face and nose padding ensures comfortable fit
- Frosted upper part protects against light reflection
- Easy headband adjustment
: EN 166, EN 170