Thianthrene, Sigma-Aldrich®
Catalog # SIAL122440-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:216.33 g/mol
:M-Clarity™ quality level = MQ100
EINECS:202-197-0
CAS Number:92-85-3
Formula:C₁₂H₈S₂
MDL Number:MFCD00005065