Order Entry
Germany
Orders LinkContactUsLinkComponent
(S)-(-)-1,1'-Bi-2-naphthol 99%

(S)-(-)-1,1'-Bi-2-naphthol 99%

Irritant
(S)-(-)-1,1'-Bi-2-naphthol 99%
Certificates
(S)-(-)-1,1'-Bi-2-naphthol 99%

3 Options Available Below

Product Details & Documents
Synonyms:  (S)-BINOL, (S)-(-)-1,1'-Binaphthyl-2,2'-Diol, (S)-[1,1'-Binaphthalene]-2,2'-diol
  • CAS Number:18531-99-2
  • Formula:C₂₀H₁₄O₂
  • MDL Number:MFCD00004068
  • Molecular Weight:286.33 g/mol
  • Melting Pt:205…211 °C
  • Storage Temperature:Ambient
  • ADR:6.1,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to light grey Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    205 to 211 °C
  • HPLC:
    ≥98.5 %
  • Specific optical rotation:
    -33° to -38° (20 °C, 589 nm) (c=1, THF)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 28,00 €
    28,00 €
    1 * 1 g
       28,00 €

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 81,20 €
    81,20 €
    1 * 5 g
       81,20 €

  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 267,00 €
    267,00 €
    1 * 25 g
       267,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.