Organic reference standard, 9,10-Anthraquinone 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA10281000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:84-65-1
- Description:9,10-Anthraquinone 100 µg/ml in acetonitrile
- Formula:C₁₄H₈O₂
- Description:9,10-Anthraquinone