Parathion (Parathion-ethyl, O,O-Diethyl O-4-nitrophenyl phosphorothioate)
Catalog # EHERC15880000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:291.26 g/mol
EINECS:200-271-7
CAS Number:56-38-2
Formula:C₁₀H₁₄NO₅PS