Dinobuton (2-sec-Butyl-4,6-dinitrophenyl isopropyl carbonate)
Catalog # EHERC12790000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:326.31 g/mol
EINECS:213-546-1
CAS Number:973-21-7
Formula:C₁₄H₁₈N₂O₇