

Diisopropyl azodicarboxylate for synthesis, Sigma-Aldrich®
8.14566.0051
: Merck
:
- Pack type:Glass bottle
- Pk:50 mL
- Seq:2370
- Solubility:(20 °C) insoluble
- Molar Mass:202.2 g/mol
- Boiling Point:75 °C (33.3 Pa)
- Vapor Pressure:1.04 hPa (20 °C)
- Flash Point:106 °C
- Density:1.03 g/cm3 (20 °C)
:
:202.21 g/mol
:9,III
:M-Clarity™ quality level = MQ200
:219-502-8
:3082
:GHS07
:GHS09
:Ambient
:2446-83-5
:C₈H₁₄N₂O₄
:MFCD00008875