Cyclopentyl methyl ether (CPME) stabilised for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:5614-37-9
:
Formula:C₆H₁₂O
MDL Number:MFCD08276401
:100.16 g/mol
Density:0.86 g/cm³ (20 °C)
Flash Pt:–1 °C (closed cup)
UN:3271
ADR:3,II
:M-Clarity™ quality level = MQ200
:GHS02
:GHS07
Specification Test Results
- Melting Point:-140 °C
- Flash Point:59.9 hPa (25 °C)
- Refractive Index:-1 °C
- Density:0.86 g/cm3 (20 °C)