Pesticide reference standard, Parathion (Parathion-ethyl, O,O-Diethyl O-4-nitrophenyl phosphorothioate) 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL15880000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:56-38-2
- Description:Parathion (Parathion-ethyl, O,O-Diethyl O-4-nitrophenyl phosphorothioate) 10 µg/ml in cyclohexane
- Formula:C₁₀H₁₄NO₅PS
- Description:Parathion (Parathion-ethyl, O,O-Diethyl O-4-nitrophenyl phosphorothioate)