Order Entry
Germany
ContactUsLinkComponent
Pesticide reference standard, Parathion (Parathion-ethyl, O,O-Diethyl O-4-nitrophenyl phosphorothioate) 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Pesticide reference standard, Parathion (Parathion-ethyl, O,O-Diethyl O-4-nitrophenyl phosphorothioate) 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL15880000AL
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, Parathion (Parathion-ethyl, O,O-Diethyl O-4-nitrophenyl phosphorothioate) 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL15880000AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    56-38-2
  • Description:
    Parathion (Parathion-ethyl, O,O-Diethyl O-4-nitrophenyl phosphorothioate) 10 µg/ml in acetonitrile
  • Formula:
    C₁₀H₁₄NO₅PS
  • Description:
    Parathion (Parathion-ethyl, O,O-Diethyl O-4-nitrophenyl phosphorothioate)

Specifications

About this item