Order Entry
Germany
ContactUsLinkComponent
 
Pesticide reference standard, Chlorobenzilate 100 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERXA11390000CY
 :  
Pesticide reference standard, Chlorobenzilate 100 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERXA11390000CY
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    510-15-6
  • Description:
    Chlorobenzilate 100 µg/ml in cyclohexane
  • Formula:
    C₁₆H₁₄Cl₂O₃
  • Description:
    Chlorobenzilate