Order Entry
Germany
ContactUsLinkComponent
Arecolinium bromide
Arecolinium bromide
Catalog # MOLE11161105-25G
Supplier:  Molekula
CAS Number:  
Arecolinium bromide
Catalog # MOLE11161105-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :236.11 g/mol
EINECS:206-087-3
UN:1544
ADR:6.1,III
CAS Number:300-08-3
Formula:C₈H₁₄BrNO₂
MDL Number:MFCD00039041