Order Entry
Germany
ContactUsLinkComponent
Ammonium hexachlororhodate(III), Reagent Grade, Sigma-Aldrich®
Ammonium hexachlororhodate(III), Reagent Grade, Sigma-Aldrich®
Catalog # SIAL206253-1G
Supplier:  Merck
CAS Number:  
Ammonium hexachlororhodate(III), Reagent Grade, Sigma-Aldrich®
Catalog # SIAL206253-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ100
 :369.74 g/mol
EINECS:239-364-2
CAS Number:15336-18-2
Formula:H₁₂Cl₆N₃Rh
MDL Number:MFCD00003394