Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Zinc D-gluconate hydrate
Zinc D-gluconate hydrate ≥97%
Zinc D-gluconate hydrate ≥97%
:
Thermo Fisher Scientific
Zinc D-gluconate hydrate ≥97%
:
Thermo Fisher Scientific
:
4468-02-4
:
:
C₁₂H₂₂O₁₄Zn·xH₂O
:
MFCD00868110
:
455.69 g/mol (anhy)
:
14,4456
:
Ambient
:
224-736-9
msds_ALFAB23022
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years