5-Hydroxy-1-tetralone, Sigma-Aldrich®
Catalog # SIAL219975-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:162.19 g/mol
:M-Clarity™ quality level = MQ100
EINECS:248-958-0
CAS Number:28315-93-7
Formula:C₁₀H₁₀O₂
MDL Number:MFCD00001693