6-Methoxy-1-tetralone 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1078-19-9
:
Formula:C₁₁H₁₂O₂
MDL Number:MFCD00001695
Boiling Pt:171 °C (11 mmHg)
Melting Pt:77…79 °C
:176.22 g/mol
Storage Temperature:Ambient
EINECS:214-078-0
Specification Test Results
- Appearance:Yellow to light brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:77 to 79 °C
- GC:≥98.5 %