Order Entry
Germany
ContactUsLinkComponent
1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone) for synthesis, Sigma-Aldrich®
1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone) for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    5 - 6 °C
  • Molar Mass:
    146.19 g/mol
  • Boiling Point:
    113 - 116 °C (8 hPa)
  • Flash Point:
    110 °C
  • Refractive Index:
    1.5685 (20 °C, 589 nm)
  • Density:
    1.10 g/cm3 (20 °C)

Specification Test Results

Related Information