1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone) 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:529-34-0
:
Formula:C₁₀H₁₀O
MDL Number:MFCD00001688
:146.19 g/mol
Boiling Pt:127 °C (13 mmHg)
Melting Pt:5…6 °C
Density:1.096 g/cm³
Flash Pt:130 °C
EINECS:208-460-6
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear yellow to amber to brown Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.5660 to 1.5710 (20°C, 589 nm)
- GC:≥97.5 %