Order Entry
Germany
ContactUsLinkComponent
Herbicide reference standard, 2,4,5-T (2,4,5-trichlorophenoxy acetic acid) 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Herbicide reference standard, 2,4,5-T (2,4,5-trichlorophenoxy acetic acid) 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA17100000AL
Supplier:  EHRENSTORFER
CAS Number:  
Herbicide reference standard, 2,4,5-T (2,4,5-trichlorophenoxy acetic acid) 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA17100000AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    93-76-5
  • Description:
    2,4,5-T (2,4,5-trichlorophenoxy acetic acid) 100 µg/ml in acetonitrile
  • Formula:
    C₈H₅Cl₃O₃
  • Description:
    2,4,5-T (2,4,5-trichlorophenoxy acetic acid)

Specifications

About this item