Herbicide reference standard, 2,4,5-T (2,4,5-trichlorophenoxy acetic acid) 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL17100000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:93-76-5
- Description:2,4,5-T (2,4,5-trichlorophenoxy acetic acid) 10 µg/ml in acetonitrile
- Formula:C₈H₅Cl₃O₃
- Description:2,4,5-T (2,4,5-trichlorophenoxy acetic acid)