Herbicide reference standard, Atrazine 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL10330000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:1912-24-9
- Description:Atrazine 10 µg/ml in cyclohexane
- Formula:C₈H₁₄ClN₅
- Description:Atrazine