Order Entry
Germany
ContactUsLinkComponent
Organic reference standard, 1,1,1-Trichloroethane 1,000 µg/ml in methanol
Organic reference standard, 1,1,1-Trichloroethane 1,000 µg/ml in methanol
Catalog # CMPLCO2A.1K.ML.1
CAS Number:  
Organic reference standard, 1,1,1-Trichloroethane 1,000 µg/ml in methanol
Catalog # CMPLCO2A.1K.ML.1
CAS Number:  

Specifications

  • Pack type:
    Glass ampoule
  • Pk:
    1 Ampoul
  • CAS No.:
    71-55-6
  • Description:
    1,1,1-Trichloroethane 1,000 µg/ml in methanol
  • Formula:
    C₂H₃Cl₃
  • Description:
    1,1,1-Trichloroethane

Specifications

About this item