Order Entry
Germany
ContactUsLinkComponent
Benfluralin
Benfluralin
Catalog # EHERC10470000
Supplier:  EHRENSTORFER
CAS Number:  
Benfluralin
Catalog # EHERC10470000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :335.28
EINECS:217-465-2
CAS Number:1861-40-1
Formula:C₁₃H₁₆F₃N₃O₄