Pesticide reference standard, Dicofol (2,2,2-Trichloro-1,1-bis(4-chlorophenyl)ethanol) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL12570000IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:115-32-2
- Description:Dicofol (2,2,2-Trichloro-1,1-bis(4-chlorophenyl)ethanol) 10 µg/ml in isooctane
- Formula:C₁₄H₉Cl₅O
- Description:Dicofol (2,2,2-Trichloro-1,1-bis(4-chlorophenyl)ethanol)