Order Entry
Germany
ContactUsLinkComponent
 
Uranium standard solution, 10 mg/L U in dil. nitric acid (from U₃O₂(NO₃)₂) Certipur® standard for ICP, Supelco®
  1.70360.0100
 :  Merck
 :  
undefined
Uranium standard solution, 10 mg/L U in dil. nitric acid (from U₃O₂(NO₃)₂) Certipur® standard for ICP, Supelco®
  1.70360.0100
 :  Merck
 :  

 

  • Concentration:
    10 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Uranium
  • Symbol:
    U

 

 

 : Traceable to SRM from NIST