
Diethyl allylmalonate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2049-80-1
:
Formula:C₁₀H₁₆O₄
MDL Number:MFCD00009155
:200.23 g/mol
Density:1.01 g/cm³
Flash Pt:92 °C
EINECS:218-072-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4300 to 1.4320 (20°C, 589 nm)
- GC:≥96.0 %
- Enantiomeric excess:≥97.5 %
- Specific optical rotation:+29° to +33° (20°C, 589 nm) (Neat)