Order Entry
Germany
ContactUsLinkComponent
Mercury standard solution, 1000 mg/L Hg in 5% nitric acid (from Hg), standard for AAS, Specpure®
Mercury standard solution, 1000 mg/L Hg in 5% nitric acid (from Hg), standard for AAS, Specpure®
Catalog # 88079.AP
CAS Number:  
Mercury standard solution, 1000 mg/L Hg in 5% nitric acid (from Hg), standard for AAS, Specpure®
Catalog # 88079.AP
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • MDL No.:
    MFCD00011035
  • Element:
    Mercury
  • Symbol:
    Hg

Specifications

About this item

Molecular Formula: Hg in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011035

Certifications: Traceable to NIST