

Chloroform : isoamyl alcohol (24:1) (v/v)
Catalog # SERA39554.01
Supplier: Serva
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
Storage Temperature:Ambient
UN:1888
ADR:6.1,III
:GHS08
:GHS07
Formula:CHCl₃,C₅H₁₂O