Order Entry
Germany
ContactUsLinkComponent
Cadmium standard solution, 5000 mg/L Cd in 1 M nitric acid (from Cd(NO₃)₂) standard for AAS
Cadmium standard solution, 5000 mg/L Cd in 1 M nitric acid (from Cd(NO₃)₂) standard for AAS
Catalog # KRAF03626.2000
Supplier:  Bernd Kraft
CAS Number:  
Cadmium standard solution, 5000 mg/L Cd in 1 M nitric acid (from Cd(NO₃)₂) standard for AAS
Catalog # KRAF03626.2000
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Concentration:
    5000 mg/L
  • Matrix:
    1 M HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Cadmium
  • Symbol:
    Cd

Specifications

About this item

Certifications: Traceable to NIST