Order Entry
Germany
ContactUsLinkComponent
 
Sodium bisulphite ≥58.5%, granular, AR® ACS, Macron Fine Chemicals™
 
undefined
Sodium bisulphite ≥58.5%, granular, AR® ACS, Macron Fine Chemicals™
 

 

  • ACS - Assay (as SO₂):
    ≥58.5 %
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Chloride (Cl):
    ≤0.02 %
  • ACS - Heavy Metals (as Pb):
    ≤0.001 %
  • ACS - Iron (Fe):
    ≤0.002 %