Order Entry
Germany
ContactUsLinkComponent
Organic reference standard, 1,1-Dichloropropene 1000 µg/ml in methanol
Organic reference standard, 1,1-Dichloropropene 1000 µg/ml in methanol
Catalog # ABCRAB144168.0001
Supplier:  abcr
CAS Number:  
Organic reference standard, 1,1-Dichloropropene 1000 µg/ml in methanol
Catalog # ABCRAB144168.0001
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    563-58-6
  • Description:
    1,1-Dichloropropene 1,000 µg/ml in methanol
  • Formula:
    C₃H₄Cl₂
  • Description:
    1,1-Dichloropropene

Specifications

About this item