Hydrochloric acid 20%
Catalog # OVER813281
Supplier: OVERLACK CHEMISCHE PRODUKTE
CAS Number:
Specifications
- Pk:10 kg
- Seq:4158
Specifications
About this item
:
ADR:8,II
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324