Potassium chloride saturated with silver chloride aqueous solution 3.5 mol/l (3.5 N) in aqueous solution
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:7447-40-7
:
Formula:KCl+AgCl
MDL Number:MFCD00011360
:74.55 g/mol
Density:1.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-211-8