Cadmium standard solution, 1000 mg/L Cd in 0,5 M nitric acid (from Cd(NO₃)₂) standard for ICP
Catalog # KRAF03828.0000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:0,5 M HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Cadmium
- Symbol:Cd
Specifications
About this item
Certifications: Traceable to NIST