Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tert-Butyl methyl ether
tert-Butyl methyl ether analytical reagent
tert-Butyl methyl ether analytical reagent
:
Bernd Kraft
tert-Butyl methyl ether analytical reagent
:
Bernd Kraft
:
1634-04-4
:
:
(CH₃)₃COCH₃
:
MFCD00008812
:
88.15 g/mol
:
13,06059
:
55.2 °C (1013 hPa)
:
–108.6 °C
:
0.7405 g/cm³ (20 °C)
:
–28 °C
:
Ambient
:
216-653-1
:
2398
:
3,II
:
GHS02
:
GHS07