Chloroform ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
366927-2.5L
:
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
- Identity (IR):complying
- Purity excluding stabilizer (GC):Min. 99.80 %
- Ethanol (GC):0.5 - 1.0 %
- Non-volatile matter:Max. 0.0003 %
- Water (Karl Fischer):Max. 0.01 %
- Free acid and chloride:complying
- Lead (Pb):Max. 0.05 ppm
- Free chlorine:complying
- Reaction against H₂SO₄:complying
- Acetone, aldehydes (neßler test):complying
- Absorbance at 400 nm:Max. 0.01
- Absorbance at 290 nm:Max. 0.01
- Absorbance at 270 nm:Max. 0.02
- Absorbance at 260 nm:Max. 0.15
- Absorbance at 255 nm:Max. 0.15
- Absorbance at 245 nm:Max. 1.00
- APHA:Max. 10
:
:119.38 g/mol
:13,02160
:62 °C
:-64 °C
:–64 °C
:1.49 g/cm³ (20 °C)
:Does not flash
: Does not flash
:Ambient
:200-663-8
:1888
:6.1,III
:Stabilised with ethanol 0.5-1.0%
:05-2114614906-45
:GHS07
:GHS08
:67-66-3
:CHCl₃
:MFCD00000826