Order Entry
Germany
Orders LinkContactUsLinkComponent
Diethyl ether stabilised analytical reagent

Diethyl ether stabilised analytical reagent

Flammable
Irritant
Supplier:   Bernd Kraft
Diethyl ether stabilised analytical reagent
Supplier:  Bernd Kraft
Certificates
Diethyl ether stabilised analytical reagent
Diethyl ether stabilised analytical reagent

5 Options Available Below

Product Details & Documents
Synonyms:  Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
  • CAS Number:60-29-7
  • Formula:(CH₃CH₂)₂O
  • MDL Number:MFCD00011646
  • Molecular Weight:74.12 g/mol
  • Merck Index:12,03852
  • Boiling Pt:35 °C (1013 hPa)
  • Melting Pt:–123 °C
  • Density:0.7135 g/cm³ (20 °C)
  • Flash Pt:–40 °C
  • UN:1155
  • ADR:3,I
  •  :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
  • Storage Temperature:Ambient
  • EINECS:200-467-2
  • REACH:01-2119535785-29
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 86,80 €
    86,80 €
    1 * 500 mL
       86,80 €

  • Glass bottle
    2.500 mL
    Log in for availability
    Log in for availability
    1 * 2.500 mL
    1 * 2.500 mL - 289,00 €
    289,00 €
    1 * 2.500 mL
       289,00 €

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 141,00 €
    141,00 €
    1 * 1 L
       141,00 €

  • 5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 480,00 €
    480,00 €
    1 * 5 L
       480,00 €

  • 25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.